HUMANPBMC-0002644
Frozen Human Normal PBMC
human peripheral blood (mononuclear cells)
Synonym(s):
Immune Cells, PBMC, Peripheral Blood Mononuclear Cells
About This Item
product name
Frozen Human Normal PBMC, 100 Million Cells/per vial
biological source
human peripheral blood (mononuclear cells)
form
frozen liquid
packaging
vial of 100 Million cells
manufacturer/tradename
BioIVT
technique(s)
cell culture | mammalian: suitable
color
cloudy
storage temp.
-140 to -196°C
General description
We are pleased to offer these products from BioIVT.
Peripheral Blood Mononuclear Cells (PBMCs) are a subset of leukocytes (or White Blood Cell, WBC), which are integral components of the body′s immune system that fight disease. PBMCs consist of a mixture of lymphocytes (T cells, B cells, and NK cells) and monocytes and are isolated from peripheral whole blood via density gradient centrifugation and red blood cell lysis.
Used for a variety of applications, PBMCs are frequently utilized in drug discovery/development, assay validation/development, and other immunology applications.
Packaging
Other Notes
Disclaimer
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service