Skip to Content
Merck
All Photos(1)

Key Documents

Y0001567

Docusate sodium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Docusate sodium salt, AOT, Bis(2-ethylhexyl) sulfosuccinate sodium salt, Sodium bis(2-ethylhexyl) sulfosuccinate, Sulfobutanedioic acid bis(2-ethylhexyl ester) sodium salt, Sulfosuccinic acid bis(2-ethylhexyl) ester sodium salt, ‘Dioctyl’ sulfosuccinate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H37NaO7S
CAS Number:
Molecular Weight:
444.56
Beilstein:
4117588
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

docusate calcium

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O

InChI

1S/C20H38O7S.Na/c1-5-9-11-16(7-3)14-26-19(21)13-18(28(23,24)25)20(22)27-15-17(8-4)12-10-6-2;/h16-18H,5-15H2,1-4H3,(H,23,24,25);/q;+1/p-1

InChI key

APSBXTVYXVQYAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Forms reverse micelles in hydrocarbon solvents; Suitable for the solubilization of the major myelin transmembrane proteolipid

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierre Paré et al.
Canadian journal of gastroenterology & hepatology, 28(10), 549-557 (2014-11-13)
Constipation is an uncomfortable and common condition that affects many, irrespective of age. Since 1500 BC and before, health care practitioners have provided treatments and prevention strategies to patients for chronic constipation despite the significant variation in both medical and
Teresa Ruston et al.
Canadian oncology nursing journal = Revue canadienne de nursing oncologique, 23(4), 236-246 (2014-01-17)
Opioid-induced constipation (OIC) is a side effect of opioid therapy that can affect quality of life, adherence to treatment, and morbidity and possibly mortality. To investigate whether docusate sodium, sennosides, and lactulose have equal efficacy and side effect profiles compared
Catalina Sandoval et al.
PloS one, 10(4), e0123669-e0123669 (2015-04-24)
Reactors found in nature can be described as micro-heterogeneous systems, where media involved in each micro-environment can behave in a markedly different way compared with the properties of the bulk solution. The presence of water molecules in micro-organized assemblies is
Anju Gupta et al.
Marine pollution bulletin, 87(1-2), 164-170 (2014-08-31)
Adsorption of suspended particles to the interface of surfactant-dispersed oil droplets can alter emulsion phase and sedimentation behavior. This work examines the effects of model mineral aggregates (silica nanoparticle aggregates or SNAs) on the behavior of oil (octane)-water emulsions prepared
Ericka Rodríguez-León et al.
Nanoscale research letters, 10, 101-101 (2015-04-18)
We have synthesized silver nanoparticles in the non-polar phase of non-aqueous microemulsions. The nanocrystals have been grown by reducing silver ions in the microemulsion cylindrical micelles formed by the reducing agent (ethylene glycol). By a careful deposit of the microemulsion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service