콘텐츠로 건너뛰기
Merck
  • Structural basis for DNA 3'-end processing by human tyrosyl-DNA phosphodiesterase 1.
Structural basis for DNA 3'-end processing by human tyrosyl-DNA phosphodiesterase 1.
Nature communications (2018-01-04)
Fiona J Flett, Emilija Ruksenaite, Lee A Armstrong, Shipra Bharati, Roberta Carloni, Elizabeth R Morris, C Logan Mackay, Heidrun Interthal, Julia M Richardson
초록
Tyrosyl-DNA phosphodiesterase (Tdp1) is a DNA 3'-end processing enzyme that repairs topoisomerase 1B-induced DNA damage. We use a new tool combining site-specific DNA-protein cross-linking with mass spectrometry to identify Tdp1 interactions with DNA. A conserved phenylalanine (F259) of Tdp1, required for efficient DNA processing in biochemical assays, cross-links to defined positions in DNA substrates. Crystal structures of Tdp1-DNA complexes capture the DNA repair machinery after 3'-end cleavage; these reveal how Tdp1 coordinates the 3'-phosphorylated product of nucleosidase activity and accommodates duplex DNA. A hydrophobic wedge splits the DNA ends, directing the scissile strand through a channel towards the active site. The F259 side-chain stacks against the -3 base pair, delimiting the junction of duplexed and melted DNA, and fixes the scissile strand in the channel. Our results explain why Tdp1 cleavage is non-processive and provide a molecular basis for DNA 3'-end processing by Tdp1.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Nuclease P1 from Penicillium citrinum, lyophilized powder, ≥200 units/mg protein (E1%/280, 3′-5′-Phosphodiesterase)