콘텐츠로 건너뛰기
Merck
  • Neuropathy-specific alterations in a Mexican population of diabetic patients.
Neuropathy-specific alterations in a Mexican population of diabetic patients.
BMC neurology (2017-08-27)
Angélica Carbajal-Ramírez, Rebeca García-Macedo, Carlos Manlio Díaz-García, Carmen Sanchez-Soto, Araceli Méndez Padrón, Jorge Escobedo de la Peña, Miguel Cruz, Marcia Hiriart
초록
Neuropathy is one of the major complications of type 2 diabetes mellitus. Our first aim was to determine the clinical characteristics of a population of diabetic patients with different types of neuropathy. Our next goal was to characterize the cytokine profile (IL-6 and IL-10), nerve growth factor (NGF) and circulating cell-adhesion molecules in these patients. Finally, we aimed to compare the renal function among the groups of neuropathic patients. In a cross-sectional study, we included 217 diabetic patients classified in three groups: sensory polyneuropathy with hypoesthesia (DS The circulating levels of NGF were significantly lower in diabetic patients, compared to healthy subjects. The range of IL-6 and IL-10 levels in neuropathic patients was higher than the control groups; however, several samples yielded null measurements. Neuropathic patients also showed increased circulating levels of the adhesion molecules ICAM, VCAM, and E-Selectin, compared to the ND group. Moreover, neuropathic patients showed reduced glomerular filtration rates compared to healthy subjects (82-103 ml/min per 1.73 m Some particular alterations in neuropathic patients included -but were not limited to- changes in circulating NGF, cell adhesion molecules, inflammation, and the worsening of the renal function. This study supports the need for further clinical surveillance and interventions considering a neuropathy-related basis.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Human NGF R  ELISA Kit, for serum, plasma, cell culture supernatant and urine