- Inwardly rectifying potassium channels influence
Inwardly rectifying potassium channels influence
Development (Cambridge, England) (2017-07-08)
Giri Raj Dahal, Sarala Joshi Pradhan, Emily Anne Bates
PMID28684627
초록
Loss of embryonic ion channel function leads to morphological defects, but the underlying reason for these defects remains elusive. Here, we show that inwardly rectifying potassium (Irk) channels regulate release of the