- TGF-β
TGF-β
American journal of physiology. Lung cellular and molecular physiology (2017-03-25)
Weichao Guo, Shigeki Saito, Cecilia G Sanchez, Yan Zhuang, Rafael E Gongora Rosero, Bin Shan, Fayong Luo, Joseph A Lasky
PMID28336812
초록
Myofibroblasts are important mediators of fibrogenesis; thus blocking fibroblast-to-myofibroblast differentiation (FMD) may be an effective strategy to treat pulmonary fibrosis (PF). Previously, we reported that histone deacetylase 4 (HDAC4) activity is necessary for transforming growth factor-β
MATERIALS