콘텐츠로 건너뛰기
Merck
  • p27(kip1) Knockout enhances collateralization in response to hindlimb ischemia.
p27(kip1) Knockout enhances collateralization in response to hindlimb ischemia.
Journal of vascular surgery (2015-02-24)
Galit Ankri-Eliahoo, Kevin Weitz, Timothy C Cox, Gale L Tang
초록
The natural response to arterial occlusive disease is enlargement of collaterals; however, the molecular factors that control collateralization are not well understood. The gene p27(Kip1) (p27) affects human response to arterial injury. Previous studies have shown that overexpression of p27 inhibits vascular endothelial and vascular smooth muscle cell (VSMC) proliferation and angiogenesis. To test the hypothesis that knockout of p27 would improve collateralization in reaction to ischemia, we performed in vivo and in vitro experiments using p27 knockout (p27(-/-)) and wild-type (wt) mice. Hindlimb ischemia was induced by left femoral artery ligation in p27(-/-) and wt (C57BL/6) female mice. The mice underwent weekly laser Doppler perfusion imaging of the footpads until sacrifice on postoperative day 28 followed by microcomputed tomography scanning of both hindlimbs. VSMCs were isolated from p27(-/-) and wt mice and used in migration and gel contraction assays in the absence and presence of the nonspecific matrix metalloproteinase (MMP) inhibitor BB94. MMP-2 and MMP-9 messenger RNA (mRNA) expression was measured by quantitative reverse transcription-polymerase chain reaction in p27(-/-) and wt VSMCs. p27(-/-) mice reperfused more effectively than wt mice by laser Doppler starting from day 7 (ischemic/nonischemic ratio, 0.33 ± 0.02 vs 0.25 ± 0.02; P < .05) and continuing through day 28 (0.45 ± 0.04 vs 0.31 ± 0.04; P < .05). The gracilis collateral diameter was similar for the nonischemic hindlimbs of the p27(-/-) and wt mice, and this collateral pathway increased similarly after ischemia as assessed by microcomputed tomography. However, the p27(-/-) mice significantly enlarged a novel collateral pathway that bridged directly between the femoral artery proximal to the ligation site and the saphenous or popliteal artery distal to the ligation site more than wt mice (158 ± 18.3 vs 82 ± 22 μm; P < .001). p27(-/-) VSMCs migrated more (79% ± 5% vs 56% ± 6%; P < .05) and caused more gel contraction (18% ± 5% of the initial area vs 43% ± 4%; P < .05) than wt cells. Migration and collagen contraction were abolished in p27(-/-) and wt cells by MMP inhibition. p27(-/-) cells expressed significantly more MMP-2 mRNA than wt cells did. Knockout of p27 enhances arterial collateralization in response to hindlimb ischemia through enlargement of a new collateral pathway. In vitro, knockout of p27 increases collagen gel contraction in addition to stimulating VSMC migration. We speculate that p27 may affect collateralization through its role in regulating MMP-2 expression.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
MISSION® esiRNA, targeting human CDKN1B