콘텐츠로 건너뛰기
Merck
  • Synthesis and structure-activity relationships of 5,6,7,8-tetrahydro-4H-thieno[3,2-b]azepine derivatives: novel arginine vasopressin antagonists.
Synthesis and structure-activity relationships of 5,6,7,8-tetrahydro-4H-thieno[3,2-b]azepine derivatives: novel arginine vasopressin antagonists.
Journal of medicinal chemistry (2003-12-30)
Hidetsura Cho, Kengo Murakami, Hiroyuki Nakanishi, Akitaka Fujisawa, Hirotaka Isoshima, Misako Niwa, Kazuhide Hayakawa, Yasunori Hase, Itsuo Uchida, Hidenori Watanabe, Korekiyo Wakitani, Kazuo Aisaka
초록
A variety of novel heterocyclic compounds having thienoazepine, pyrroloazepine, furoazepine, and thienodiazepine skeletons were synthesized, most of which exhibited potent antagonism of [(3)H]-AVP specific binding in assays using rat liver (V1), rat kidney (V2), human platelet plasma membranes, and recombinant human CHO cells (V2), as well as antagonizing AVP-induced hypertension in rats (V1, intravenous) and showing a diuretic effect in rats (V2, oral). By detailed studies of the structure-activity relationships of these compounds, the thienoazepine derivative 1 was found to be a very potent combined V1 and V2 antagonist. After further pharmacological and toxicological evaluation as well as physical properties, the hydrochloride 2 (JTV-605) of compound 1 was selected for clinical studies as a potent AVP antagonist with a long duration of action.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
4-Piperidinopiperidine, 97%