콘텐츠로 건너뛰기
Merck
  • Direct-acting antiviral-based triple therapy on alpha-fetoprotein level in chronic hepatitis C patients.
Direct-acting antiviral-based triple therapy on alpha-fetoprotein level in chronic hepatitis C patients.
World journal of gastroenterology (2015-04-29)
Koji Takayama, Norihiro Furusyo, Eiichi Ogawa, Hiroaki Ikezaki, Motohiro Shimizu, Masayuki Murata, Jun Hayashi
초록
To investigate the impact of telaprevir-based triple therapy on the serum alpha-fetoprotein (AFP) level of chronic hepatitis C patients. A total of 210 patients with chronic hepatitis C genotype 1 of high viral load (baseline serum hepatitis C virus RNA > 5.0 log10 IU/mL) were divided into two groups by type of treatment: triple therapy with telaprevir, pegylated-interferon-α (PEG-IFNα), and ribavirin (RBV) for 24 wk (n = 88), or dual therapy with PEG-IFNα and RBV for 48 wk (n = 122). The relationship between virological response and the change in the serum AFP level from baseline to 24 wk after the end of treatment was examined. No significant difference in mean baseline AFP level was found between the triple and dual therapy groups (8.8 ng/mL vs 7.8 ng/mL). Triple therapy produced significant declines in the AFP level in sustained virological response (SVR) and non-SVR patients (7.8 ng/mL at baseline to 3.5 ng/mL at 24 wk after the end of treatment, P < 0.001 and 14.3 ng/mL to 9.5 ng/mL, P = 0.004, respectively). In contrast, dual therapy resulted in a significant decline in AFP level only in SVR patients (4.7 ng/mL to 2.8 ng/mL, P < 0.001), but not in non-SVR patients (10.2 ng/mL to 10.1 ng/mL). Among patients with a high-baseline AFP level (≥ 10 ng/mL), the decline in the AFP level was significantly higher in the triple therapy than in the dual therapy group (15.9 ng/mL vs 1.6 ng/mL, P = 0.037). Regardless of virological response, telaprevir-based triple therapy reduced the serum AFP level.