콘텐츠로 건너뛰기
Merck
  • The biogenetic anatomy of vitamin B6. A 13C NMR investigation of the biosynthesis of pyridoxol in Escherichia coli.
The biogenetic anatomy of vitamin B6. A 13C NMR investigation of the biosynthesis of pyridoxol in Escherichia coli.
The Journal of biological chemistry (1996-11-29)
R E Hill, K Himmeldirk, I A Kennedy, R M Pauloski, B G Sayer, E Wolf, I D Spenser
초록
It is shown by incorporation experiments with 13C bond-labeled substrates, followed by analysis by means of 13C NMR spectroscopy, that two compounds, 1-deoxy-D-xylulose (12) and 4-hydroxy-L-threonine (), serve as precursors of pyridoxol (vitamin B6) (1) in Escherichia coli. Together, these two compounds account for the entire C8N skeleton of the vitamin. 1-Deoxy-D-xylulose supplies the intact C5 unit, C-2',2,3,4,4' of pyridoxol. 4-Hydroxy-L-threonine undergoes decarboxylation in supplying the intact C3N unit, N-1,C-6,5,5'. Both precursors are ultimately derived from glucose. The C5 unit of pyridoxol that is derived from 1-deoxy-D-xylulose originates by union of a triose phosphate (yielding C-3,4,4') with pyruvic acid (which decarboxylates to yield C-2',2). D-Erythroate (11) enters the C3 unit, C-6,5,5', and is therefore an intermediate on the route from glucose into 4-hydroxy-L-threonine.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
D-Erythronic acid potassium salt, ≥97.0% (GC)