콘텐츠로 건너뛰기
Merck
  • Sacral plexus injury after radiotherapy for carcinoma of cervix.
Sacral plexus injury after radiotherapy for carcinoma of cervix.
Cancer (1990-10-01)
J A Stryker, K Sommerville, R Perez, D E Velkley
초록
A 42-year-old woman developed lower extremity weakness and sensory loss 1 year after external and intracavitary radiotherapy for Stage IB carcinoma of cervix. She has been followed for 5 years posttreatment, and the neurologic abnormalities have persisted, but no evidence of recurrent carcinoma has been found. We believe this to be a rare case of sacral plexus radiculopathy developing as a late complication after radiotherapy. Suggestions are made for improving the radiotherapy technique to prevent this complication in future cases.