콘텐츠로 건너뛰기
Merck
  • LUMA interacts with emerin and influences its distribution at the inner nuclear membrane.
LUMA interacts with emerin and influences its distribution at the inner nuclear membrane.
Journal of cell science (2008-01-31)
Luiza Bengtsson, Henning Otto
초록
We present here a first characterization of LUMA, an unique integral inner nuclear membrane (INM) protein. LUMA is a highly conserved protein even in some bacteria and shares a PFAM domain of unknown function with orthologs from many species. Assessing LUMA topology by using protease protection of membrane-inserted LUMA and antibody epitope accessibility assays reveals that LUMA contains four transmembrane domains and a large hydrophilic domain located between membrane spans 1 and 2. The large hydrophilic domain is exposed to the perinuclear space whereas both LUMA termini reside cyto- or nucleoplasmically. Nuclear envelope targeting of LUMA mainly depends on the membrane spans. LUMA's transmembrane domains also promote homooligomerization. LUMA binds A- and B-type lamins and depends on A-type lamins for its INM localization. Furthermore, it interacts with emerin. Both downregulation of LUMA and overexpression of dominant-negative acting LUMA fragments causes redistribution of emerin. We propose that LUMA functions as a tetraspanin-like membrane organizer and has the potential to contribute to the pathomechanism of dystrophic diseases, such as Emery-Dreifuss muscular dystrophy.