- Upstream-binding factor is sequestered into herpes simplex virus type 1 replication compartments.
Upstream-binding factor is sequestered into herpes simplex virus type 1 replication compartments.
The Journal of general virology (2008-12-18)
Nigel D Stow, Vanessa C Evans, David A Matthews
PMID19088274
초록
Previous reports have shown that adenovirus recruits nucleolar protein upstream-binding factor (UBF) into adenovirus DNA replication centres. Here, we report that despite having a different mode of viral DNA replication, herpes simplex virus type 1 (HSV-1) also recruits UBF into viral DNA replication centres. Moreover, as with adenovirus, enhanced green fluorescent protein-tagged fusion proteins of UBF inhibit viral DNA replication. We propose that UBF is recruited to the replication compartments to aid replication of HSV-1 DNA. In addition, this is a further example of the role of nucleolar components in viral life cycles.