콘텐츠로 건너뛰기
Merck
  • 2,6-Dimethylheptanoyl-CoA is a specific substrate for long-chain acyl-CoA dehydrogenase (LCAD): evidence for a major role of LCAD in branched-chain fatty acid oxidation.
2,6-Dimethylheptanoyl-CoA is a specific substrate for long-chain acyl-CoA dehydrogenase (LCAD): evidence for a major role of LCAD in branched-chain fatty acid oxidation.
Biochimica et biophysica acta (1998-08-26)
R J Wanders, S Denis, J P Ruiter, L IJlst, G Dacremont
초록
Oxidation of straight-chain fatty acids in mitochondria involves the complicated interaction between a large variety of different enzymes. So far four different mitochondrial straight-chain acyl-CoA dehydrogenases have been identified. The physiological function of three of the four acyl-CoA dehydrogenases has been resolved in recent years especially from studies on patients suffering from certain inborn errors of mitochondrial fatty acid beta-oxidation. The physiological role of long-chain acyl-CoA dehydrogenase (LCAD) has remained obscure, however. The results described in this paper provide strong evidence suggesting that LCAD plays a central role in branched-chain fatty acid metabolism since it turns out to be the major acyl-CoA dehydrogenase reacting with 2,6-dimethylheptanoyl-CoA, a metabolite of pristanic acid, which itself is the alpha-oxidation product of phytanic acid.