콘텐츠로 건너뛰기
Merck
Long coiled-coil proteins and membrane traffic.
Biochimica et biophysica acta (2003-08-14)
Alison K Gillingham, Sean Munro
초록
Protein transport between organelles is mediated by vesicles which must accurately dock and fuse with appropriate compartments. Over the past several years a large number of long coiled-coil proteins have been identified on the Golgi and on endosomes, mostly as auto-antigens in autoimmune disorders. Based on their restricted intracellular distributions and their predicted rod-like structure, these proteins have been proposed to play a role in tethering vesicles to target organelles prior to fusion. However, such proteins may also play a structural role, for example as components of a Golgi matrix, or as scaffolds for the assembly of other factors important for fusion. This review will examine what is known about the function of these large coiled-coil proteins in membrane traffic.