콘텐츠로 건너뛰기
Merck
  • Genomic structure, alternative maturation and tissue expression of the human BBOX1 gene.
Genomic structure, alternative maturation and tissue expression of the human BBOX1 gene.
Biochimica et biophysica acta (2006-11-18)
Caroline Rigault, Françoise Le Borgne, Jean Demarquoy
초록
Gamma-butyrobetaine hydroxylase (BBOX1) is the enzyme responsible for the biosynthesis of l-carnitine, a key molecule of fatty acid metabolism. This cytosolic dimeric protein belongs to the dioxygenase family. In human, enzyme activity has been detected in kidney, liver and brain. The human gene encoding gamma-butyrobetaine hydroxylase is located on chromosome 11. Although the protein structure and activity have been extensively described, little information is available concerning BBOX1 structure and expression. In this study, the organization of the human gene was determined. The structure and functions of the 5'- and 3'-untranslated regions of the human BBOX1 mRNA were characterized in kidney, liver and brain. Our experiments revealed that the transcription initiation of the human BBOX1 gene might occur at 3 different exons, and that the expression level of each type of transcript is organ-specific. We showed that the use of 3 different promoters is responsible for the 5'-end heterogeneity. Investigations on BBOX1 mRNA maturation highlighted an alternative polyadenylation mechanism that generates two 3'-untranslated regions differing by their length. This alternative polyadenylation exhibited a tissue specificity.