콘텐츠로 건너뛰기
Merck
  • Cannabinoid receptor 2 is critical for the homing and retention of marginal zone B lineage cells and for efficient T-independent immune responses.
Cannabinoid receptor 2 is critical for the homing and retention of marginal zone B lineage cells and for efficient T-independent immune responses.
Journal of immunology (Baltimore, Md. : 1950) (2011-11-04)
Sreemanti Basu, Avijit Ray, Bonnie N Dittel
초록
The endocannabinoid system has emerged as an important regulator of immune responses, with the cannabinoid receptor 2 (CB2) and its principle ligand 2-archidonoylglycerol playing a major role. How CB2 regulates B cell functions is not clear, even though they express the highest levels of CB2 among immune cell subsets. In this study, we show that CB2-deficient mice have a significant reduction in the absolute number of marginal zone (MZ) B cells and their immediate precursor, transitional-2 MZ precursor. The loss of MZ lineage cells in CB2(-/-) mice was shown to be B cell intrinsic using bone marrow chimeras and was not due to a developmental or functional defect as determined by B cell phenotype, proliferation, and Ig production. Furthermore, CB2(-/-) B cells were similar to wild type in their apoptosis, cell turnover, and BCR and Notch-2 signaling. We then demonstrated that CB2(-/-) MZ lineage B cells were less efficient at homing to the MZ and that their subsequent retention was also regulated by CB2. CB2(-/-) mice immunized with T-independent Ags produced significantly less Ag-specific IgM. This study demonstrates that CB2 positively regulates T-independent immune responses by controlling the localization and positioning of MZ lineage cells to the MZ.