- Synthesis of dixanthones and poly(dixanthone)s by cyclization of 2-aryloxybenzonitriles in trifluoromethanesulfonic acid.
Synthesis of dixanthones and poly(dixanthone)s by cyclization of 2-aryloxybenzonitriles in trifluoromethanesulfonic acid.
Organic letters (2001-07-21)
H M Colquhoun, D F Lewis, D J Williams
PMID11463310
초록
[reaction: see text] Condensation of 2-fluorobenzonitriles with phenoxides affords 2-aryloxybenzonitriles that cyclize cleanly in trifluoromethanesulfonic acid at room temperature to give xanthone-iminium triflates. The C=N bond in these compounds is remarkably resistant to hydrolysis, but prolonged reaction with strong aqueous acid under vigorous conditions affords xanthones in good yield. The synthesis is exemplified for a novel series of polynuclear dixanthones and for a high molar mass polyxanthone derived from the previously unreported monomer 3,3'-difluoro-4,4'-biphenyldicarbonitrile.
MATERIALS