콘텐츠로 건너뛰기
Merck
  • Synthesis of Succinimido[3,4-b]indane and 1,2,3,4,5,6-Hexahydro-1,5-methano-3-benzazocine-2,4-dione by Sequential Alkylation and Intramolecular Arylation of Enolates Derived from N,N,N'N'-Tetramethylbutanediamides and N,N,N'N'-Tetramethylpentanediamides.
Synthesis of Succinimido[3,4-b]indane and 1,2,3,4,5,6-Hexahydro-1,5-methano-3-benzazocine-2,4-dione by Sequential Alkylation and Intramolecular Arylation of Enolates Derived from N,N,N'N'-Tetramethylbutanediamides and N,N,N'N'-Tetramethylpentanediamides.
The Journal of organic chemistry (2001-10-25)
Sushama A. Dandekar, Stacey N. Greenwood, Thomas D. Greenwood, Stéphane Mabic, Joseph S. Merola, James M. Tanko, James F. Wolfe
초록
The tricyclic title compounds, 4 and 5, were synthesized by initial alkylation of the lithium monoenolates of N,N,N',N'-tetramethylbutanediamide (6) and N,N,N',N'-tetramethylpentanediamide (19) with 2-iodobenzyl chloride in liquid NH(3) at -60 degrees C to afford 2-(2-iodobenzyl)-N,N,N',N'-tetramethylbutanediamide (9) and 2-(2-iodobenzyl)-N,N,N',N'-tetramethylpentanediamide (20) in yields of 88 and 87%, respectively. Treatment of 9 with KNH(2) in liquid NH(3) resulted in formation and intramolecular arylation of the less-substituted alpha-enolate to produce trans-1,2-bis(N,N-dimethylcarboxamido)indane (10a) in 60% yield. Selective hydrolysis of 10a with aqueous Na(2)O(2) gave trans-1-(N,N-dimethylcarboxamido)indane-2-carboxylic acid (17), which was then converted to bridged succinimide 4 by transformation to trans-1-(N,N-dimethylcarboxamido)indane-2-carboxamide (10c) followed by cyclization of this mixed primary/tertiary amide by means of NaH in refluxing THF. Treatment of 20 with KNH(2) in liquid NH(3) led to intramolecular arylation and accompanying ammonolysis to afford trans-1-(N,N-dimethylcarboxamido)-1,2,3,4-tetrahydronaphthalene-3-carboxamide (21b). Conversion of 21b to 5 was similarly effected by means of NaH. Experiments designed to test the mechanistic aspects of the intramolecular arylations provided evidence for competing aryne and SET pathways.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
2-Iodobenzyl chloride, 98%