콘텐츠로 건너뛰기
Merck
  • Metabolism of isomeric nitrobenzo[a]pyrenes leading to DNA adducts and mutagenesis.
Metabolism of isomeric nitrobenzo[a]pyrenes leading to DNA adducts and mutagenesis.
Mutation research (1997-05-12)
P P Fu, F Y Qui, H Jung, L S Von Tungeln, D J Zhan, M J Lee, Y S Wu, R H Heflich
초록
We have been interested in determining the structural and electronic features that may be useful in predicting the mutagenic activity of nitro-polycyclic aromatic hydrocarbons (nitro-PAHs). We have previously found that a correlation between structural and electronic features and direct-acting mutagenicity in Salmonella typhimurium cannot be made using nitro-PAHs with different molecular size. In this study, a series of structurally related nitro-PAHs, the environmental contaminants 1-, 3-, and 6-nitrobenzo[alpha]pyrene (NBaP) and their derivatives, was used to determine structure-activity relationships. It was found that isomeric NBaPs are activated to DNA damaging and mutagenic derivatives by nitroreduction, ring-oxidation, or by a combination of these two pathways. A general finding was that NBaPs and derivatives with their nitro substituent oriented perpendicular to the aromatic system exhibit either very weak or no direct-acting mutagenicity in S. typhimurium strains TA98 and TA100. In this paper, we also discuss the effect of the location of the nitro group on the metabolism and the mutagenicity of NBaPs and the effect of oxygen-containing functional groups on the mutagenicity of NBaP derivatives. These findings provide a useful molecular basis for interpreting and predicting the direct-acting mutagenicity of nitro-PAHs.
MATERIALS
제품 번호
브랜드
제품 설명
6-Nitrobenzo[a]pyrene, BCR®, certified reference material