콘텐츠로 건너뛰기
Merck
  • A urinary metabolite ratio that reflects systemic caffeine clearance.
A urinary metabolite ratio that reflects systemic caffeine clearance.
Clinical pharmacology and therapeutics (1987-08-01)
M E Campbell, S P Spielberg, W Kalow
초록
Systemic caffeine clearance and urinary metabolite profiles were determined in 15 subjects with diverse exposure histories to cytochrome P-450 inducers (cigarette smoke) and inhibitors (oral contraceptive steroids). A correlation was observed between caffeine clearance and a urinary ratio based on the molar recovery of paraxanthine 7-demethylation products relative to a paraxanthine 8-hydroxylation product (r = 0.91; P less than 0.001). Analysis of urinary metabolites was undertaken in a larger population to assess the effects of gender, age, oral contraceptives, and smoking on the ratio. No gender differences were observed in either adults or children; children (n = 21) showed a higher (P less than 0.001) mean metabolite ratio than adults (n = 61), oral contraceptive users (n = 9) had lower (P less than 0.05) ratios than women not taking oral contraceptives (n = 30), and smokers (n = 26) had higher (P less than 0.001) ratios than nonsmokers (n = 61). The data indicate that a urinary metabolite ratio based on paraxanthine 7-demethylation/8-hydroxylation products reflects systemic caffeine clearance and likely monitors cytochrome P-450 activity inducible by polycyclic aromatic hydrocarbons.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
1,7-Dimethyluric acid, ≥97.0% (HPLC)