콘텐츠로 건너뛰기
Merck
  • Pyrazoloquinoline benzodiazepine receptor ligands: effects on schedule-controlled behavior in dogs.
Pyrazoloquinoline benzodiazepine receptor ligands: effects on schedule-controlled behavior in dogs.
Pharmacology, biochemistry, and behavior (1985-08-01)
H E Shannon, W A Thompson
초록
The effects of diazepam and the pyrazoloquinoline benzodiazepine receptor ligands CGS8216, CGS9896, and CGS9895 on schedule-controlled responding were studied in dogs. Responding was maintained under a multiple fixed-interval (FI) 5-min fixed-ratio (FR) 30 response schedule of food presentation. Diazepam (PO) produced dose-related decreases in response rates under FR component. Under the FI, rates first increased and then decreased with increasing doses of diazepam. Diazepam also produced a dose-related disruption of the temporal pattern of responding under the FI as measured by decreases in quarter-life values. CGS8216 IV produced dose-related decreases in response rates under both components. The highest oral dose of CGS8216 also decreased rates in both components. CGS8216 was approximately 100 times more potent by the IV route as compared to the oral route. CGS9896 IV had no significant effect on responding under either component of the multiple schedule. However, with increasing doses of CGS9896 PO, response rates under both components first decreased and then returned to control values. CGS9895 PO was without significant effect on responding. When CGS8216 was administered concomitantly with graded doses of diazepam, the former drug blocked the rate-decreasing effects of diazepam under the FR component, but not the rate-increasing effects of diazepam under the FI. The present results demonstrate that although these three pyrazoloquinolines are benzodiazepine receptor ligands, they do not exhibit diazepam-like effects on schedule-controlled behavior.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
CGS 9895, ≥98% (HPLC)