콘텐츠로 건너뛰기
Merck
  • Powerful vascular protection by combining cilnidipine with valsartan in stroke-prone, spontaneously hypertensive rats.
Powerful vascular protection by combining cilnidipine with valsartan in stroke-prone, spontaneously hypertensive rats.
Hypertension research : official journal of the Japanese Society of Hypertension (2012-11-30)
Shinji Takai, Denan Jin, Shizuka Aritomi, Kazumi Niinuma, Mizuo Miyazaki
초록
Cilnidipine is an L- and N-type calcium channel blocker (CCB), and amlodipine is an L-type CCB. Valsartan (10 mg kg(-1)), valsartan (10 mg kg(-1)) and amlodipine (1 mg kg(-1)), and valsartan (10 mg kg(-1)) and cilnidipine (1 mg kg(-1)) were administered once daily for 2 weeks to stroke-prone, spontaneously hypertensive rats (SHR-SPs). Blood pressure was significantly reduced by valsartan, and it was further reduced by the combination therapies. Vascular endothelial dysfunction was significantly attenuated in all therapeutic groups, and further significant attenuation was observed in the valsartan+cilnidipine-treated group, but not in the valsartan+amlodipine-treated group. Vascular nicotinamide adenine dinucleotide phosphate (NADPH) oxidase subunit NOX1 gene expression was significantly attenuated in all therapeutic groups, and significantly greater attenuation was observed in the valsartan+cilnidipine-treated group than in the valsartan-treated group. Compared with the valsartan-treated group, the positive areas for 4-hydroxy-2-nonenal were significantly lower only in the valsartan+cilnidipine-treated group. Plasma renin activity was significantly augmented in the valsartan-treated group, and it was significantly attenuated in the valsartan+cilnidipine-treated group. A significant increase in the ratio of plasma angiotensin-(1-7) to angiotensin II was observed only in the valsartan+cilnidipine-treated group. Vascular angiotensin-converting enzyme (ACE) gene expression was significantly attenuated only in the valsartan+cilnidipine-treated group, but ACE2 gene expression was significantly higher in all of the therapeutic groups. Thus, valsartan and cilnidipine combination therapy might have a powerful protective effect in the vascular tissues via increases in the angiotensin-(1-7)/angiotensin II ratio in plasma.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Cilnidipine, ≥98% (HPLC), powder