콘텐츠로 건너뛰기
Merck
  • Isocyanide binding to the copper(I) centers of the catalytic core of peptidylglycine monooxygenase (PHMcc).
Isocyanide binding to the copper(I) centers of the catalytic core of peptidylglycine monooxygenase (PHMcc).
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2001-07-27)
F C Rhames, N N Murthy, K D Karlin, N J Blackburn
초록
Binding of the Cu(I)-specific ligands 2,6-dimethylphenyl isocyanide (DIMPI) and isopropyl isocyanide (IPI) to the reduced form of peptidylglycine monooxygenase (PHM) is reported. Both ligands bind to the methionine-containing CuM center, eliciting FTIR bands at 2,138 and 2,174 cm(-1), respectively, but appear unable to coordinate at the histidine-containing CuH center in the wild-type enzyme. This chemistry parallels that previously observed for CO binding to the reduced PHM catalytic core (PHMcc). However, in contrast to the CO chemistry, peptide substrate binding did not induce binding of the isocyanide at CuH. XAS confirmed the binding of DIMPI at CuM via the observation of a short Cu-C interaction at 1.87 A and by the lengthening of the Cu-S(methionine) bond length by 0.06 A. Similarly, FTIR studies on DIMPI binding to the M314I and H172A mutant forms of reduced PHMcc confirmed the assignment of the 2,138-cm(-1) IR band as a CuM-DIMPI complex, but surprisingly also showed DIMPI binding to CuH, as indicated by a band at 2,148 cm(-1). An inorganic complex, [Cu(1,2-Me2Im)2(DIMPI)](PF6), was synthesized and its crystal structure was determined as a model for the interaction of isocyanides with imidazole-containing Cu(I) complexes. Comparison of EXAFS data for the protein and model suggests that DIMPI probably binds to CuM in a tilted fashion, similar to that of ethyl isocyanide binding to myoglobin.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Isopropyl isocyanide, 97%