콘텐츠로 건너뛰기
Merck
  • Treatment of ametryn in wastewater by a hybrid MBR system: a lab-scale study.
Treatment of ametryn in wastewater by a hybrid MBR system: a lab-scale study.
Water science and technology : a journal of the International Association on Water Pollution Research (2012-07-26)
D Navaratna, L Shu, K Baskaran, V Jegatheesan
초록
Agricultural discharge of herbicides to the Great Barrier Reef (GBR) poses significant threat to the marine ecosystem. This study evaluates the performance of a hybrid treatment system consists of a membrane bioreactor (MBR), UV disinfection unit and a granular activated carbon (GAC) column in treating ametryn which is one of the major herbicides in agricultural discharges. While the MBR alone removes only 40% of ametryn at a hydraulic retention time of 7.8 h, the hybrid system removed ametryn to below detection levels.
MATERIALS
제품 번호
브랜드
제품 설명
Supelco
Ametryn, PESTANAL®, analytical standard