콘텐츠로 건너뛰기
Merck
  • Weak estrogenic transcriptional activities of Bisphenol A and Bisphenol S.
Weak estrogenic transcriptional activities of Bisphenol A and Bisphenol S.
Toxicology in vitro : an international journal published in association with BIBRA (2012-04-18)
Elise Grignard, Silvia Lapenna, Susanne Bremer
초록
In 2011, the European Commission has restricted the use of Bisphenol A in plastic infant feeding bottles. In a response to this restriction, Bisphenol S is now often used as a component of plastic substitutes for the production of babybottles. One of the major concerns leading to the restriction of Bisphenol A was its weak estrogenic activity. By using two highly standardised transactivation assays, we could demonstrate that the estrogenic activity of Bisphenol A and Bisphenol S is of a comparable potency. Furthermore, some insights about the structure-activity relationships of these two chemicals and their metabolites could be gained from in silico predictions of their relative estrogen receptor-binding affinities and their liver phase-I biotransformation.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
4,4′-Sulfonyldiphenol, 98%