콘텐츠로 건너뛰기
Merck
  • Comparative genomics and functional analysis of the NiaP family uncover nicotinate transporters from bacteria, plants, and mammals.
Comparative genomics and functional analysis of the NiaP family uncover nicotinate transporters from bacteria, plants, and mammals.
Functional & integrative genomics (2011-09-29)
Linda Jeanguenin, Aurora Lara-Núñez, Dmitry A Rodionov, Andrei L Osterman, Nataliya Y Komarova, Doris Rentsch, Jesse F Gregory, Andrew D Hanson
초록
The transporter(s) that mediate uptake of nicotinate and its N-methyl derivative trigonelline are not known in plants, and certain mammalian nicotinate transporters also remain unidentified. Potential candidates for these missing transporters include proteins from the ubiquitous NiaP family. In bacteria, niaP genes often belong to NAD-related regulons, and genetic evidence supports a role for Bacillus subtilis and Acinetobacter baumannii NiaP proteins in uptake of nicotinate or nicotinamide. Other bacterial niaP genes are, however, not in NAD-related regulons but cluster on the chromosome with choline-related (e.g., Ralstonia solanacearum and Burkholderia xenovorans) or thiamin-related (e.g., Thermus thermophilus) genes, implying that they might encode transporters for these compounds. Radiometric uptake assays using Lactococcus lactis cells expressing NiaP proteins showed that B. subtilis, R. solanacearum, and B. xenovorans NiaP transport nicotinate via an energy-dependent mechanism. Likewise, NiaP proteins from maize (GRMZM2G381453, GRMZM2G066801, and GRMZM2G081774), Arabidopsis (At3g13050), and mouse (SVOP) transported nicotinate; the Arabidopsis protein also transported trigonelline. In contrast, T. thermophilus NiaP transported only thiamin. None of the proteins tested transported choline or the thiazole and pyrimidine products of thiamin breakdown. The maize and Arabidopsis NiaP proteins are the first nicotinate transporters reported in plants, the Arabidopsis protein is the first trigonelline transporter, and mouse SVOP appears to represent a novel type of mammalian nicotinate transporter. More generally, these results indicate that specificity for nicotinate is conserved widely, but not absolutely, among pro- and eukaryotic NiaP family proteins.
MATERIALS
제품 번호
브랜드
제품 설명
Supelco
Trigonelline hydrochloride, analytical standard
제품번호
팩 크기
재고 정보
가격
수량