콘텐츠로 건너뛰기
Merck
  • [Enzyme immunoassay for the determination of hexestrol in meat].
[Enzyme immunoassay for the determination of hexestrol in meat].
Prikladnaia biokhimiia i mikrobiologiia (2011-03-30)
M M Vdovenko, C F Peng, C L Xu, E S Vylegzhanina, A A Komarov, I Iu Sakharov
초록
An indirect competitive enzyme-linked immunosorbent assay (ELISA) of hexestrol (HES), an antibiotic forbidden for use in livestock farming, has been developed. Conditions of ELISA have been optimized by varying the concentrations of the coating conjugate (HES-ovalbumin), anti-HES antiserum, casein, and Tween 20. In the absence of Tween 20 in the reaction mixture, the detection limit (IC10) equaled 0.01 ng/ml, IC50 equaled 0.17 ng/ml, and the working range (IC20-IC80) equaled 0.03-0.86 ng/ml, while, in the presence of 0.05% Tween 20, these values equaled 0.05 ng/ml, 2.9 ng/ml, and 0.26-32.0 ng/ml, respectively. Standard deviation of the analysis results did not exceed 5.4%. If ELISA was performed in the absence of detergents, the recovery value upon HES determination in spiked beef samples ranged from 74 to 147%.