콘텐츠로 건너뛰기
Merck
  • Early changes in gene expression induced by tobacco smoke: Evidence for the importance of estrogen within lung tissue.
Early changes in gene expression induced by tobacco smoke: Evidence for the importance of estrogen within lung tissue.
Cancer prevention research (Philadelphia, Pa.) (2010-06-03)
Sibele I Meireles, Gustavo H Esteves, Roberto Hirata, Suraj Peri, Karthik Devarajan, Michael Slifker, Stacy L Mosier, Jing Peng, Manicka V Vadhanam, Harrell E Hurst, E Jordao Neves, Luiz F Reis, C Gary Gairola, Ramesh C Gupta, Margie L Clapper
초록
Lung cancer is the leading cause of cancer deaths in the United States, surpassing breast cancer as the primary cause of cancer-related mortality in women. The goal of the present study was to identify early molecular changes in the lung induced by exposure to tobacco smoke and thus identify potential targets for chemoprevention. Female A/J mice were exposed to either tobacco smoke or HEPA-filtered air via a whole-body exposure chamber (6 h/d, 5 d/wk for 3, 8, and 20 weeks). Gene expression profiles of lung tissue from control and smoke-exposed animals were established using a 15K cDNA microarray. Cytochrome P450 1b1, a phase I enzyme involved in both the metabolism of xenobiotics and the 4-hydroxylation of 17beta-estradiol (E(2)), was modulated to the greatest extent following smoke exposure. A panel of 10 genes were found to be differentially expressed in control and smoke-exposed lung tissues at 3, 8, and 20 weeks (P < 0.001). The interaction network of these differentially expressed genes revealed new pathways modulated by short-term smoke exposure, including estrogen metabolism. In addition, E(2) was detected within murine lung tissue by gas chromatography-coupled mass spectrometry and immunohistochemistry. Identification of the early molecular events that contribute to lung tumor formation is anticipated to lead to the development of promising targeted chemopreventive therapies. In conclusion, the presence of E(2) within lung tissue when combined with the modulation of cytochrome P450 1b1 and other estrogen metabolism genes by tobacco smoke provides novel insight into a possible role for estrogens in lung cancer.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
4-Hydroxyestradiol