- Semisynthesis and antiproliferative evaluation of a series of 3'-aminoflavones.
Semisynthesis and antiproliferative evaluation of a series of 3'-aminoflavones.
Bioorganic & medicinal chemistry letters (2009-05-22)
Jérôme Quintin, Didier Buisson, Sylviane Thoret, Thierry Cresteil, Guy Lewin
PMID19457664
초록
A series of 3'-aminoflavones 5,6,7,8-tetra- or 5,7-dioxygenated on the A-ring was synthesized from tangeretin or naringin, two natural Citrus flavonoids. These flavones were evaluated for antiproliferative activity, activation of apoptosis, and inhibition of tubulin assembly. The most antiproliferative flavones exhibit a common 5-hydroxy-6,7,8-trimethoxy substitution pattern on the A-ring.
MATERIALS