콘텐츠로 건너뛰기
Merck
  • Parallel solution-phase synthesis of 4H-benzo[1,4]thiazin-3-one and 1,1-dioxo-1,4-dihydro-2H-1lambda6-benzo[1,4]thiazin-3-one derivatives from 1,5-difluoro-2,4-dinitrobenzene.
Parallel solution-phase synthesis of 4H-benzo[1,4]thiazin-3-one and 1,1-dioxo-1,4-dihydro-2H-1lambda6-benzo[1,4]thiazin-3-one derivatives from 1,5-difluoro-2,4-dinitrobenzene.
Journal of combinatorial chemistry (2007-05-17)
Zhanguo Wang, Yunyun Yuan, Yuyan Chen, Gangchun Sun, Xianghong Wu, Shengmei Zhang, Chunyan Han, Guanxing Wang, Li Li, Gang Liu
초록
This paper discusses the synthesis of privileged structures 4H-benzo[1,4]thiazin-3-one and 1,1-dioxo-1,4-dihydro-2H-1lambda6-benzo[1,4]thiazin-3-one derivatives in a parallel solution-phase manner using 1,5-difluoro-2,4-dinitrobenzene. Each scaffold possesses four diversity points. A cheap and efficient oxidant, urea-hydrogen peroxide (UHP), was applied for the introduction of the sulfone group. The intramolecular cyclization to 1,1-dioxo-1,4-dihydro-2H-1lambda6-benzo[1,4]thiazin-3-one was achieved by microwave assistance or the use of an inorganic base.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
1,5-Difluoro-2,4-dinitrobenzene, 97%