콘텐츠로 건너뛰기
Merck
  • Advantageous application of a surface coil to EPR irradiation in overhauser-enhanced MRI.
Advantageous application of a surface coil to EPR irradiation in overhauser-enhanced MRI.
Magnetic resonance in medicine (2007-03-29)
Shingo Matsumoto, Kenichi Yamada, Hiroshi Hirata, Keiji Yasukawa, Fuminori Hyodo, Kazuhiro Ichikawa, Hideo Utsumi
초록
The present study describes the advantageous application of a surface coil to electron paramagnetic resonance (EPR) irradiation in Overhauser-enhanced MRI (OMRI). OMRI is a double-resonance method for imaging free radicals based on the Overhauser effect. Proton NMR images are recorded without and with EPR irradiation of the free radical resonance, which results in a difference proton image that shows signal enhancement in spatial regions that contain the free radical. To obtain good signal enhancement in OMRI, very high RF power and a long EPR irradiation time are required. To improve sensitivity and shorten the image acquisition time, especially for localized (and topical) applications, we developed and tested a surface-coil-type EPR irradiation coil. Theoretical calculations and experimental data showed that EPR irradiation through the surface coil could ameliorate the localized Overhauser enhancement, which was related to the ratio of B(1) surface coil/B(1) volume coil in the region of interest (ROI), as expected. The increased sensitivity could also be converted into a shortened EPR irradiation time, resulting in fast data acquisition. For biomedical applications, the use of a surface coil (as opposed to a conventional volume coil) could decrease the total RF power deposition in the sample required to obtain the same Overhauser enhancement in the ROI.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
3-Carbamoyl-PROXYL