콘텐츠로 건너뛰기
Merck
  • (Sub)-picosecond spectral evolution of fluorescence in photoactive proteins studied with a synchroscan streak camera system.
(Sub)-picosecond spectral evolution of fluorescence in photoactive proteins studied with a synchroscan streak camera system.
Photochemistry and photobiology (2006-04-15)
I H M van Stokkum, B Gobets, T Gensch, F van Mourik, K J Hellingwerf, R van Grondelle, J T M Kennis
초록
The spectral evolution of three photoactive proteins has been investigated by measuring the fluorescence with good temporal and wavelength resolution and a high signal-to-noise ratio. Upon excitation at 400 nm wild-type (wt) PYP both at neutral pH and in the low-pH blueshifted pBdark state exhibited a strong quenching of the fluorescence, the major part of which could be described by lifetimes of about 1.7 and 7.7 ps. The remaining fluorescence decay occurred multiexponentially with lifetimes between 30 and 125 ps. Additionally, in wtPYP at neutral pH, a dynamic Stokes shift was found to occur with a time constant of about 0.25 ps. In a PYP preparation that was reconstituted with the chromophore 7-hydroxy-coumarin-3- carboxylic acid rather than the native coumaric acid, and which is therefore not capable of performing the cis-trans-isomerization that initiates the photocycle in wtPYP, the fluorescence was found to decay multiexponentially with lifetimes of 51 ps, 0.33 and 3.77 ns. Additionally, dynamic Stokes shifts were observed with time constants of about 0.1 and 3.5 ps. Upon comparison of the dynamics of this preparation with that of wtPYP the multiexponential decay with lifetimes of 1.7 and 7.7 ps found in wtPYP was attributed to photochemistry of the p-coumaric-acid chromophore. The emission from bacteriorhodopsin mutant D85S upon excitation at 635 nm decays biexponentially with estimated lifetimes of 5.2 and 19.1 ps. No dynamic Stokes shift was observed here. Four lifetimes were needed to describe the decay of the emission from the A* state in the green fluorescent protein. From a target analysis it was concluded that the longer lifetimes are accompanied by a decreasing probability of forming I*, which approaches zero with the longest A* lifetime of 1.5 ns. These observations may be explained by heterogeneity of A and by relaxation of A*. In all three systems studied, multiexponential decay of emission was present, suggesting that heterogeneity is a common feature of these chromophore protein complexes.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Coumarin-3-carboxylic acid, 99%