콘텐츠로 건너뛰기
Merck
  • A new fluorescent chemosensor for copper ions based on tripeptide glycyl-histidyl-lysine (GHK).
A new fluorescent chemosensor for copper ions based on tripeptide glycyl-histidyl-lysine (GHK).
Organic letters (2001-10-12)
Y Zheng, Q Huo, P Kele, F M Andreopoulos, S M Pham, R M Leblanc
초록
[structure: see text]. A new fluorescent chemosensor for Cu2+ ions was synthesized by modifying the tripeptide glycyl-histidyl-lysine (GHK) with 9-carbonylanthracene via the standard Fmoc solid-phase peptide synthesis method. While significant fluorescence quenching was observed from the molecule upon binding with Cu2+, addition of Fe2+, Co2+, Ni2+, and Zn2+ to the peptide solution caused a minimum fluorescence emission spectral change, indicating a high specificity of this chemosensor for Cu2+ ions. Effects of pH were also investigated.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Gly-His-Lys acetate salt, BioXtra, γ-irradiated, suitable for cell culture
Sigma-Aldrich
Gly-His-Lys acetate salt, ≥97% (TLC)