콘텐츠로 건너뛰기
Merck
  • Characterization of the binding of a potent, selective, radioiodinated antagonist to the human neurokinin-1 receptor.
Characterization of the binding of a potent, selective, radioiodinated antagonist to the human neurokinin-1 receptor.
Molecular pharmacology (1992-09-01)
M A Cascieri, E Ber, T M Fong, S Sadowski, A Bansal, C Swain, E Seward, B Frances, D Burns, C D Strader
초록
We have synthesized a potent, selective, radioiodinated antagonist of the human neurokinin-1 (NK1) receptor and have characterized its binding to the cloned receptor expressed in Chinese hamster ovary cells. (cis)-2-(Diphenylmethyl)-N-[(2-iodophenyl)-methyl]-1- azabicyclo[2.2.2]octan-3-amine (L-703606) inhibits binding of 125I-Tyr8-substance P to the human NK1 receptor with an IC50 of 2 nM. This compound is a competitive antagonist of substance P-induced inositol phosphate generation, with a Kb of 29 nM. [125I]L-703606 binds to a single class of high affinity binding sites in human NK1/Chinese hamster ovary cell membranes (Kd = 0.3 nM). Substance P inhibits the binding of [125I]L-703606 to 65% of the NK1 receptor sites with a Kd of 0.04 +/- 0.03 nM and to the remaining 35% of the sites with a Kd of 1.5 +/- 0.7 nM. Addition of the nonhydrolyzable GTP analog guanylyl-5'-(beta, gamma-imido)diphosphate [Gpp(NH)p] shifts greater than 90% of the binding sites to the lower affinity state. In addition, Gpp(NH)p markedly alters the dissociation of substance P from the NK1 receptor by increasing the number of sites in the low affinity, rapidly dissociating state. However, Gpp(NH)p does not affect the rate of dissociation of [125I]L-703606. These data suggest that the pharmacological properties of [125I]L-703606 binding to the human NK1 receptor are similar to those of antagonists of nonpeptide guanine nucleotide-binding protein-coupled receptors and that this ligand will be useful for the biochemical and pharmacological characterization of the human NK1 receptor.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
L-703,606 oxalate salt hydrate, solid