콘텐츠로 건너뛰기
Merck
Molecular probes for extracellular adenosine receptors.
Biochemical pharmacology (1987-05-15)
K A Jacobson, D Ukena, W Padgett, K L Kirk, J W Daly
초록
Derivatives of adenosine receptor agonists (N6-phenyladenosines) and antagonists (1,3-dialkyl-8-phenylxanthines) bearing functionalized chains suitable for attachment to other molecules have been reported [Jacobson et al., J. med. Chem. 28, 1334 and 1341 (1985)]. The "functionalized congener" approach has been extended to the synthesis of spectroscopic and other probes for adenosine receptors that retain high affinity (Ki approximately 10(-9)-10(-8) M) in A1-receptor binding. The probes have been synthesized from an antagonist xanthine amine congener (XAC) and an adenosine amine congener (ADAC). [3H]ADAC has been synthesized and found to bind highly specifically to A1-adenosine receptors of rat and calf cerebral cortical membranes with KD values of 1.4 and 0.34 nM respectively. The higher affinity in the bovine brain, seen also with many of the probes derived from ADAC and XAC, is associated with phenyl substituents. The spectroscopic probes contain a reporter group attached at a distal site of the functionalized chain. These bifunctional ligands may contain a spin label (e.g. the nitroxyl radical TEMPO) for electron spin resonance spectroscopy, or a fluorescent dye, including fluorescein and 4-nitrobenz-2-oxa-1,3-diazole (NBD), or labels for 19F nuclear magnetic resonance spectroscopy. Potential applications of the spectroscopic probes in characterization of adenosine receptors are discussed.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Fmoc-D-Lys(Biotin)-OH, ≥95%