콘텐츠로 건너뛰기
Merck
  • Membrane perturbing properties of natural phenolic and resorcinolic lipids.
Membrane perturbing properties of natural phenolic and resorcinolic lipids.
FEBS letters (2008-10-07)
Maria Stasiuk, Arkadiusz Kozubek
초록
The effects induced by natural phenolic and resorcinolic lipids on membrane permeability were investigated. All of the compounds tested perturbed the phospholipid bilayer and stabilized erythrocytes against hypoosmotically induced hemolysis. Dipalmitoylphosphatidylcholine liposomes with two preincorporated fluorescent dyes (1-(4-trimethylammoniumphenyl)-6-phenyl-1,3,5-hexatrien p-toluenesulfonate (TMA-DPH) and N-(-nitrobenz-2-oxa-1,3-diazol-4-yl)-1,2-dihexadecanoyl-sn-glycero-3-phosphoetanolamine triethylammonium salt (NBD-PE)) were used to determine the effects of tested compounds on the core and surface of the bilayer. Resorcinolic lipids from rye and cardol increased the polarization of TMA-DPH fluorescence more than that of NBD-PE, but anacardic acid, methylocardol, and alkylphenol increased NBD-PE dye fluorescence.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
N,N,N-Trimethyl-4-(6-phenyl-1,3,5-hexatrien-1-yl)phenylammonium p-toluenesulfonate, BioReagent, suitable for fluorescence, ≥96.0% (TLC)