콘텐츠로 건너뛰기
Merck
  • A2B adenosine receptor activation and modulation by protein kinase C.
A2B adenosine receptor activation and modulation by protein kinase C.
iScience (2023-07-05)
Zhan-Guo Gao, Ian M Levitan, Asuka Inoue, Qiang Wei, Kenneth A Jacobson
초록
Protein kinase C (PKC) isoforms regulate many important signaling pathways. Here, we report that PKC activation by phorbol 12-myristate 13-acetate (PMA) enhanced A2B adenosine receptor (AR)-mediated, but not β2-adrenergic receptor-mediated, cAMP accumulation, in H9C2 cardiomyocyte-like and HEK293 cells. In addition to enhancement, PKC (PMA-treatment) also activated A2BAR with low Emax (H9C2 and NIH3T3 cells endogenously expressing A2BAR), or with high Emax (A2BAR-overexpressing HEK293 cells) to induce cAMP accumulation. A2BAR activation induced by PKC was inhibited by A2BAR and PKC inhibitors but enhanced by A2BAR overexpression. Gαi isoforms and PKCγ isoform were found to be involved in both enhancement of A2BAR function and A2BAR activation. Thus, we establish PKC as an endogenous modulator and activator of A2BAR, involving Giα and PKCγ. Depending on signaling pathway, PKC could activate and enhance, or alternatively inhibit A2BAR activity. These findings are relevant to common functions of A2BAR and PKC, e.g. cardioprotection and cancer progression/treatment.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
MRS 1754 hydrate, ≥98% (HPLC), solid
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)