콘텐츠로 건너뛰기
Merck
  • Cod Residual Protein Prevented Blood Pressure Increase in Zucker fa/fa Rats, Possibly by Inhibiting Activities of Angiotensin-Converting Enzyme and Renin.
Cod Residual Protein Prevented Blood Pressure Increase in Zucker fa/fa Rats, Possibly by Inhibiting Activities of Angiotensin-Converting Enzyme and Renin.
Nutrients (2018-11-25)
Iselin Vildmyren, Aslaug Drotningsvik, Åge Oterhals, Ola Ween, Alfred Halstensen, Oddrun Anita Gudbrandsen
초록
Hypertension is the leading risk factor for cardiovascular disease, and prevention of high blood pressure through diet and lifestyle should be a preferred approach. High intake of fish is associated with lower blood pressure, possibly mediated through the proteins since peptides with angiotensin-converting enzyme (ACE) inhibiting capacities have been identified in fish skin, backbone, and fillet. The effects of cod meals made from residual materials and fillet on blood pressure were investigated in obese Zucker fa/fa rats which spontaneously develop high blood pressure. Rats were fed diets containing water-soluble (stickwater) or water-insoluble (presscake) fractions of protein-rich meals from cod residual materials (head, gut, backbone with muscle residuals, skin, trimmings) or fillet. Rats were fed diets containing 25% of total protein from cod meal and 75% of protein from casein, or casein as the sole protein source (control group) for four weeks. Results show that a diet containing residual presscake meal with high gut content prevented blood pressure increase, and this cod residual meal also showed the strongest in vitro inhibitions of ACE and renin activities. In conclusion, a diet containing water-insoluble proteins (presscake meal) with high gut content prevented increase in blood pressure in obese Zucker fa/fa rats.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Nitrite/Nitrate Assay Kit, colorimetric, used for detection of nitric oxide metabolites
Sigma-Aldrich
Renin Assay Kit, sufficient for 100 fluorometric tests