콘텐츠로 건너뛰기
Merck
  • MACC1-AS1 promotes hepatocellular carcinoma cell invasion and proliferation by regulating PAX8.
MACC1-AS1 promotes hepatocellular carcinoma cell invasion and proliferation by regulating PAX8.
Aging (2020-01-10)
Hui Tong, Xiaohui Liu, Tao Li, Weihua Qiu, Chenghong Peng, Baiyong Shen, Zhecheng Zhu
초록
Long noncoding RNAs play vital roles in several biological processes, including cell growth and embryonic development. We showed that MACC1-AS1 was overexpressed in hepatocellular carcinoma (HCC) cells and tissues. The MACC1-AS1 expression level was dramatically upregulated in HCC samples compared to adjacent normal samples, and 77.5% (31 of 40) of HCC samples showed overexpression of MACC1-AS1. Ectopic MACC1-AS1 expression enhanced cell proliferation and cyclin D1 expression in both SMMC7721 and MHCC-97H cells. Ectopic expression of MACC1-AS1 promoted vimentin, N-cadherin and snail expression and decreased E-cadherin expression in both SMMC7721 and MHCC-97H cells. MACC1-AS1 overexpression also induced cell invasion in the same two cell lines. Furthermore, MACC1-AS1 overexpression enhanced PAX8 expression in HCC cells. The PAX8 level was dramatically increased in HCC samples compared to adjacent normal samples, and 75% (30 of 40) of HCC samples showed overexpression of PAX8. PAX8 expression was positively correlated with MACC1-AS1 expression in HCC samples. MACC1-AS1 overexpression promoted HCC cell proliferation, EMT and invasion through regulating PAX8. These results suggest that MACC1-AS1 acts as an oncogene in the development of HCC.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
MISSION® esiRNA, targeting human PAX8