콘텐츠로 건너뛰기
Merck
  • D1 dopamine receptors intrinsic activity and functional selectivity affect working memory in prefrontal cortex.
D1 dopamine receptors intrinsic activity and functional selectivity affect working memory in prefrontal cortex.
Molecular psychiatry (2018-12-12)
Yang Yang, Sang-Min Lee, Fumiaki Imamura, Krishne Gowda, Shantu Amin, Richard B Mailman
초록
Dopamine D1 agonists enhance cognition, but the role of different signaling pathways (e.g., cAMP or β-arrestin) is unclear. The current study compared 2-methyldihydrexidine and CY208,243, drugs with different degrees of both D1 intrinsic activity and functional selectivity. 2-Methyldihydrexidine is a full agonist at adenylate cyclase and a super-agonist at β-arrestin recruitment, whereas CY208,243 has relatively high intrinsic activity at adenylate cyclase, but much lower at β-arrestin recruitment. Both drugs decreased, albeit in dissimilar ways, the firing rate of neurons in prefrontal cortex sensitive to outcome-related aspects of a working memory task. 2-Methyldihydrexidine was superior to CY208,243 in prospectively enhancing similarity and retrospectively distinguishing differences between correct and error outcomes based on firing rates, enhancing the micro-network measured by oscillations of spikes and local field potentials, and improving behavioral performance. This study is the first to examine how ligand signaling bias affects both behavioral and neurophysiological endpoints in the intact animal. The data show that maximal enhancement of cognition via D1 activation occurred with a pattern of signaling that involved full unbiased intrinsic activity, or agonists with high β-arrestin activity.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
SCH-39166 hydrobromide, ≥98% (HPLC)