์ฝ˜ํ…์ธ ๋กœ ๊ฑด๋„ˆ๋›ฐ๊ธฐ
Merck
ADAM10 and BACE1 are localized to synaptic vesicles.
Journal of neurochemistry (2015-08-25)
Jolanta L Lundgren, Saheeb Ahmed, Sophia Schedin-Weiss, Gunnar K Gouras, Bengt Winblad, Lars O Tjernberg, Susanne Frykman
์ดˆ๋ก
Synaptic degeneration and accumulation of the neurotoxic amyloid ฮฒ-peptide (Aฮฒ) in the brain are hallmarks of Alzheimer disease. Aฮฒ is produced by sequential cleavage of the amyloid precursor protein (APP), by the ฮฒ-secretase ฮฒ-site APP cleaving enzyme 1 (BACE1) and ฮณ-secretase. However, Aฮฒ generation is precluded if APP is cleaved by the ฮฑ-secretase ADAM10 instead of BACE1. We have previously shown that Aฮฒ can be produced locally at the synapse. To study the synaptic localization of the APP processing enzymes we used western blotting to demonstrate that, compared to total brain homogenate, ADAM10 and BACE1 were greatly enriched in synaptic vesicles isolated from rat brain using controlled-pore glass chromatography, whereas Presenilin1 was the only enriched component of the ฮณ-secretase complex. Moreover, we detected ADAM10 activity in synaptic vesicles and enrichment of the intermediate APP-C-terminal fragments (APP-CTFs). We confirmed the western blotting findings using in situ proximity ligation assay to demonstrate close proximity of ADAM10 and BACE1 with the synaptic vesicle marker synaptophysin in intact mouse primary hippocampal neurons. In contrast, only sparse co-localization of active ฮณ-secretase and synaptophysin was detected. These results indicate that the first step of APP processing occurs in synaptic vesicles whereas the final step is more likely to take place elsewhere.
MATERIALS
์ œํ’ˆ ๋ฒˆํ˜ธ
๋ธŒ๋žœ๋“œ
์ œํ’ˆ ์„ค๋ช…
Sigma-Aldrich
Anti-Nicastrin Antibody, ascites fluid, clone 9C3, Chemiconยฎ