콘텐츠로 건너뛰기
Merck
  • A-769662 protects osteoblasts from hydrogen dioxide-induced apoptosis through activating of AMP-activated protein kinase (AMPK).
A-769662 protects osteoblasts from hydrogen dioxide-induced apoptosis through activating of AMP-activated protein kinase (AMPK).
International journal of molecular sciences (2014-06-25)
Yalong Zhu, Jianhua Zhou, Rongguang Ao, Baoqing Yu
초록
Here we report that 5'-monophosphate (AMP)-activated protein kinase (AMPK) agonist A-769662 inhibited hydrogen peroxide (H₂O₂)-induced viability loss and apoptosis of human and mouse osteoblast cells. H₂O₂-induced moderate AMPK activation in osteoblast cells, which was enhanced by A-769662. Inactivation of AMPK by its inhibitor compound C, or by target shRNA-mediated silencing and kinase dead (KD) mutation exacerbated H₂O₂-induced cytotoxicity in osteoblast cells. A-769662-mediated protective effect against H₂O₂ was also blocked by AMPK inhibition or depletion. A-769662 inhibited reactive oxygen species (ROS) accumulation by H₂O₂ in osteoblast cells. Meanwhile, H₂O₂-induced ATP depletion was inhibited by A-769662, but was aggravated by compound C. Further, H₂O₂ induced AMPK-dependent and pro-survival autophagy in cultured osteoblast cells, which was enhanced by A-769662. Our results suggested that activation of AMPK by H₂O₂ is anti-apoptosis and pro-survival in osteoblast cells, probably due to its anti-oxidant, pro-autophagy and ATP preservation abilities, and A-769662-mediated cell-protective effect in osteoblast cells requires AMPK activation. Our study suggests that A-769662 might be further investigated as a novel anti-osteonecrosis agent.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
A-769662, ≥98% (HPLC)