콘텐츠로 건너뛰기
Merck
  • Numb Depletion Promotes Drp1-Mediated Mitochondrial Fission and Exacerbates Mitochondrial Fragmentation and Dysfunction in Acute Kidney Injury.
Numb Depletion Promotes Drp1-Mediated Mitochondrial Fission and Exacerbates Mitochondrial Fragmentation and Dysfunction in Acute Kidney Injury.
Antioxidants & redox signaling (2018-06-13)
Ze Liu, Hao Li, Jianqun Su, Shihui Xu, Fengxin Zhu, Jun Ai, Zheng Hu, Miaomiao Zhou, Jianwei Tian, Zhiyuan Su, Peiliang Yang, Jing Nie
초록
Mitochondrial fragmentation is a crucial mechanism contributing to tubular cell apoptosis during acute kidney injury (AKI). However, the mechanism of modulating mitochondrial dynamics during AKI remains unclear. Numb is a multifunction adaptor protein that is expressed in renal tubules. The aim of the present study was to evaluate the role of Numb in mitochondrial dysfunction during AKI. The expression of Numb was upregulated in both ischemia-reperfusion- and cisplatin-induced AKI. Depletion of Numb from proximal tubules (PT-Nb-KO) exacerbated AKI shown as more severe renal tubular damage and higher serum creatinine than wild-type mice. Numb depletion alone significantly increased mitochondrial fragmentation without altering mitochondrial mass and function, including adenosine triphosphate production, mitochondrial membrane potential, oxygen consumption, and reactive oxygen species production. However, mitochondrial fragmentation and dysfunction were significantly aggravated after cisplatin exposure in PT-Nb-KO mice. Mechanistically, Numb depletion triggered dynamin-related protein 1 (Drp1) recruitment to mitochondria by increasing the phosphorylation of Drp1 at serine 656 residue (human Drp1 ser637). Inhibiting the activity of Rho-associated coiled-coil containing protein kinase (ROCK) by Y-27632 attenuated phosphorylation of Drp1 ser656 and mitochondrial fragmentation in Numb-deficient cells. Administration of mdivi-1, a pharmacological inhibitor of Drp1, restored mitochondrial morphology, attenuated cisplatin-induced tubular injury, and renal dysfunction in PT-Nb-KO mice. Innovation and Conclusion: Our data suggest that Numb depletion promotes mitochondrial fragmentation by promoting the phosphorylation of Drp1 Ser637 and thus exacerbates cisplatin-induced mitochondrial dysfunction and tubular cell apoptosis. These findings add a novel insight into modulating mechanism of mitochondrial dynamics during AKI.