콘텐츠로 건너뛰기
Merck
  • Hydrothermally tailor-made chitosan fiber for micro-solid phase extraction of petroleum acids in crude oils.
Hydrothermally tailor-made chitosan fiber for micro-solid phase extraction of petroleum acids in crude oils.
Journal of chromatography. A (2018-06-11)
Gang-Tian Zhu, Xiao-Li Hu, Sheng He, Xiao-Mei He, Shu-Kui Zhu, Yu-Qi Feng
초록
Tailor-made chitosan fiber was prepared via hydrothermal treatment to serve as a micro-solid phase extraction (micro-SPE) sorbent for the analysis of petroleum acids (PAs) in crude oils. Chitosan fiber, which is commercial and cheap, has a diameter of about 10 μm and a length of a few centimeters. The fibrous property of the sorbent enables the micro-SPE to deal with viscous crude oil samples because of the low back-pressure during extraction, while the abundant hydroxyl groups and amino groups on the surface of chitosan fiber can provide high density of specific sites for adsorption of PAs. Moreover, it was found that hydrothermal treatment at certain conditions could tune the surface properties of chitosan fiber, leading to significant improvement of the capacity of the fiber in adsorption of PAs. Using hydrothermally treated chitosan fiber as sorbent, the micro-SPE was applied to the determination of PAs in crude oils, with the advantages of easy-operation, rapidness and high sensitivity (the limits of detection range from 0.7 ng/g to 5.4 ng/g). Furthermore, coupled with comprehensive two dimensional gas chromatography-mass spectrometry (GC × GCMS), the treated chitosan fiber packed micro-SPE method showed a great potential for comprehensive profiling of PAs in crude oils.