콘텐츠로 건너뛰기
Merck
  • Assessment of cytotoxic and genotoxic effects of enniatin-A in vitro.
Assessment of cytotoxic and genotoxic effects of enniatin-A in vitro.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2018-06-12)
Sevcan Mamur, Deniz Yuzbasioglu, Serkan Yılmaz, Esra Erikel, Fatma Unal
초록
Enniatin A (EN-A) is a Fusarium mycotoxin which is a common contaminant in grains and especially in maize and it causes serious loss of product. The aim of this study was to investigate the cytotoxic effects using 3-(4,5-dimethylthiazolyl-2)-2,5 diphenyltetrazolium bromide (MTT) assay in human cervix carcinoma (HeLa) cell line, and genotoxic effects of EN-A using chromosome aberrations (CAs), sister chromatid exchanges (SCEs), micronuclei (MN) and comet assays in human lymphocytes. The cells were treated with 0.07, 0.14, 0.29, 0.57, 1.15, 2.29, 4.59 and 9.17 μM concentrations of EN-A. It exhibited cytotoxic effects in HeLa cell lines especially when the concentrations were increased. The half-inhibitory value (IC50) was determined as 1.15 μM concentration for 24 h and 0.57 μM concentration for 48 h. However, EN-A failed to affect the frequency of CAs, SCEs and MN in human lymphocytes. Only a slight increase was observed in the frequency of SCEs at 0.57 μM concentration over 48 h. The replication (RI) and nuclear division (NDI) indices were not affected. On the contrary, EN-A decreased the mitotic index (MI) significantly at all concentrations compared to the negative control and solvent control (except at 0.29 μM for 24 h, and except at 0.14, 0.29 and 0.57 μM for 48 h). Treatments over 2.29 μM showed toxic effects in human lymphocytes. EN-A significantly increased comet tail intensity (except at 0.07 and 0.57 μM) in isolated human lymphocytes. The results of this study demonstrate that EN-A has an obvious cytotoxic effect especially when the EN-A concentration was increased. In addition, EN-A could exhibit a mild genotoxic effect.