콘텐츠로 건너뛰기
Merck
  • Gold nano-flowers (Au NFs) modified screen-printed carbon electrode electrochemical biosensor for label-free and quantitative detection of glycated hemoglobin.
Gold nano-flowers (Au NFs) modified screen-printed carbon electrode electrochemical biosensor for label-free and quantitative detection of glycated hemoglobin.
Talanta (2019-05-28)
Xiao Wang, Jing Su, Dongdong Zeng, Gang Liu, Lizhuang Liu, Yi Xu, Chenguang Wang, Xinxin Liu, Lu Wang, Xianqiang Mi
초록
Glycated hemoglobin (HbA1c) represents the average glucose level over the past three months and has been considered as the most important biomarker for the diagnosis of Type Ⅱ diabetes (T2D). Herein, a label-free and quantitative electrochemical biosensor based on 4-mercaptophenylboronic acid (4-MPBA) modified gold nano-flowers (Au NFs) substrate was developed for the determination of HbA1c. Under optimal conditions, the linear dynamic ranges of HbA1c (5 μg/mL - 1000 μg/mL) and HbA1c% (2%-20%) by cyclic voltammetry were achieved. The electrochemical biosensor showed great detection specificity towards HbA1c and relatively stability after storage at 4 °C. This method could also be applied in human serum system which holds great potential to be applied to monitor real blood samples of diabetes patients. In human serum system, the recovery rate could reach 103.8% and 99.0%. It could achieve fast detection, the total analysis time was less than 65 min, and the detection time was less than 10 s. Moreover, in terms of fabrication process, operation procedure, detection time and cost, this technique was superior to the current HbA1c detection methods suggesting great promise for the practical clinical use in the future.