콘텐츠로 건너뛰기
Merck
  • Steroidogenic potential of human fetal kidney at early gestational age.
Steroidogenic potential of human fetal kidney at early gestational age.
Steroids (2019-06-01)
I Savchuk, M L Morvan, J P Antignac, M Kurek, B Le Bizec, O Söder, K Svechnikov
초록
Steroidogenic potential of the human fetal kidney (hFK) at the end of first trimester is poorly investigated. Little is known about the ontogeny of steroidogenic enzymes and activities of steroidogenic pathways in the hFK at early pregnancy. Our aim was to explore steroidogenesis and the expression of steroidogenic enzymes in the hFK at gestational weeks (GW) 9-12. Steroids in the hFK were analyzed by gas chromatography/coupled to tandem mass spectrometry. The expression of steroidogenic enzymes in the hFK at GW 9-12 was investigated by qPCR, automated Western blotting and immunohistochemistry. We observed that the hFK produced substantial amount of steroids of the Δ5 and Δ4 pathways and several steroid precursors in the biosynthesis of DHT via the backdoor pathway but not DHT itself. The levels of steroids and expression of relevant steroidogenic enzymes (e.g., CYP17A1, HSD3B1, HSD3B2, CYP11B1 and AKR1C4) we significantly higher in the hFK at GW11-12 compared to GW9. We also found the expression of sex steroid receptors (e.g., AR, ERα and ERβ) in the hFK at GW9-12. No sex-dependent differences in the levels of all identified steroids and expression of steroidogenic enzymes in the hFK from male and female fetuses were found. Altogether, our data indicate that the hFK at early pregnancy is steroidogenic organ with potential to synthesize multiple steroids that may play an important role in the formation and development of this organ in humans.