콘텐츠로 건너뛰기
Merck
  • Rhynchophylline from Uncaria rhynchophylla functionally turns delayed rectifiers into A-Type K+ channels.
Rhynchophylline from Uncaria rhynchophylla functionally turns delayed rectifiers into A-Type K+ channels.
Journal of natural products (2009-04-01)
Chun-Hsiao Chou, Chi-Li Gong, Chia-Chia Chao, Chia-Huei Lin, Chiu-Yin Kwan, Ching-Liang Hsieh, Yuk-Man Leung
초록
Rhynchophylline (1), a neuroprotective agent isolated from the traditional Chinese medicinal herb Uncaria rhynchophylla, was shown to affect voltage-gated K(+) (Kv) channel slow inactivation in mouse neuroblastoma N2A cells. Extracellular 1 (30 microM) accelerated the slow decay of Kv currents and shifted the steady-state inactivation curve to the left. Intracellular dialysis of 1 did not accelerate the slow current decay, suggesting that this compound acts extracellularly. In addition, the percent blockage of Kv currents by this substance was independent of the degree of depolarization and the intracellular K(+) concentration. Therefore, 1 did not appear to directly block the outer channel pore, with the results obtained suggesting that it drastically accelerated Kv channel slow inactivation. Interestingly, 1 also shifted the activation curve to the left. This alkaloid also strongly accelerated slow inactivation and caused a left shift of the activation curve of Kv1.2 channels heterologously expressed in HEK293 cells. Thus, this compound functionally turned delayed rectifiers into A-type K(+) channels.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Rhynchophylline, ≥95% (HPLC)