콘텐츠로 건너뛰기
Merck
  • A novel somatic mutation of SIN3A detected in breast cancer by whole-exome sequencing enhances cell proliferation through ERα expression.
A novel somatic mutation of SIN3A detected in breast cancer by whole-exome sequencing enhances cell proliferation through ERα expression.
Scientific reports (2018-10-31)
Kenji Watanabe, Shigeru Yamamoto, Syuiti Sakaguti, Keishiro Isayama, Masaaki Oka, Hiroaki Nagano, Yoichi Mizukami
초록
Breast cancer is the most frequent tumor in women, and in nearly two-thirds of cases, the tumors express estrogen receptor α (ERα, encoded by ESR1). Here, we performed whole-exome sequencing of 16 breast cancer tissues classified according to ESR1 expression and 12 samples of whole blood, and detected 310 somatic mutations in cancer tissues with high levels of ESR1 expression. Of the somatic mutations validated by a different deep sequencer, a novel nonsense somatic mutation, c.2830 C>T; p.Gln944*, in transcriptional regulator switch-independent 3 family member A (SIN3A) was detected in breast cancer of a patient. Part of the mutant protein localized in the cytoplasm in contrast to the nuclear localization of ERα, and induced a significant increase in ESR1 mRNA. The SIN3A mutation obviously enhanced MCF7 cell proliferation. In tissue sections from the breast cancer patient with the SIN3A c.2830 C>T mutation, cytoplasmic SIN3A localization was detected within the tumor regions where nuclear enlargement was observed. The reduction in SIN3A mRNA correlates with the recurrence of ER-positive breast cancers on Kaplan-Meier plots. These observations reveal that the SIN3A mutation has lost its transcriptional repression function due to its cytoplasmic localization, and that this repression may contribute to the progression of breast cancer.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
MISSION® esiRNA, targeting human ESR1